Learn More
Cesium hydroxide, 50% w/w aq. soln., 99.9% (metals basis)
Description
Cesium hydroxide is used in electric storage batteries. Also used to make porous cesium titanosilicates and potential ion-exchange materials for cleanup of radioactive wastes. Cesium Hydroxide is used as electrolyte in alkaline batteries at subzero temperatures, color photography and as a catalyst in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 25 g |
UN Number | UN2681 |
Solubility Information | Highly soluble in water. |
Formula Weight | 149.91 |
Percent Purity | 99.9% |
Concentration or Composition (by Analyte or Components) | 50% w/w aq. soln. |
Odor | Odorless |
Sensitivity | Air Sensitive |
Show More |
Safety and Handling
Hazard Category | H302-H314-H335-H361 |
Hazard Statement | GHS H Statement H314-H302 Causes severe skin burns and eye damage. Harmful if swallowed. |
Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CAESIUM HYDROXIDE SOLUTION |
EINECSNumber | 244-344-1 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.