Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium hydroxide monohydrate, 96%; cesium carbonate <5%

Catalog No. AAB2248022
Click to view available options
Quantity:
5 g
25 g
100 g
500 g

CAS: 35103-79-8 | CsH3O2 | 167.93 g/mol

Cesium hydroxide monohydrate can be used as a strong electrolyte for the battery and it also can be used as polymerization catalyst. It is also used as a raw materials for manufacturing various cesium salts and Catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cesium hydroxide monohydrate can be used as a strong electrolyte for the battery and it also can be used as polymerization catalyst. It is also used as a raw materials for manufacturing various cesium salts and Catalyst.

Solubility
Soluble in water with exothermic reaction.

Notes
Air sensitive & hygroscopic. Incompatible with oxidizing agents, acids, air, water/moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35103-79-8
Molecular Formula CsH3O2
Molecular Weight (g/mol) 167.93
MDL Number MFCD00149664
InChI Key ABSOMGPQFXJESQ-UHFFFAOYSA-M
Synonym cesium hydroxide monohydrate, cesium hydroxide hydrate, cesium hydroxide, monohydrate, cesium hydroxide-hydrate, cesium hydroxide, hydrate, ksc493c5l, caesium 1+ ion hydrate hydroxide, cesium hydroxide,monohydrate 9ci
PubChem CID 23679066
SMILES O.[OH-].[Cs+]

Specifications

Melting Point 205°C to 206°C
Quantity 100 g
UN Number UN3262
Merck Index 14,2013
Solubility Information Soluble in water with exothermic reaction.
IUPAC Name cesium;hydroxide;hydrate
Formula Weight 167.93 (149.91 Anhydrous)
Percent Purity 96%
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Density 1.87
Assay cesium carbonate <5%
Chemical Name or Material Cesium hydroxide monohydrate
Show More Show Less
Hazard Category H302-H314
Hazard Statement GHS H Statement
H314-H302
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, BASIC, INORGANIC, N.O.S.
EINECSNumber 244-344-1
RTECSNumber FK9800000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.