Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium iodide, 99.999% (metals basis)

Catalog No. p-7048599
Click to view available options
Quantity:
25 g
100 g

CAS: 7789-17-5 | CsI | 259.81 g/mol

Cesium iodide is used as an input phosphor in x-ray image intensifier tube of fluoroscopy equipment. It is also used as an equipment for electromagnetic calorimetry in experimental particle physics. It is often employed as a beamsplitter in Fourier Transform Infrared (FT-IR) spectrometers. Cesium iodide is an inorganic scintillator which is mainly used in experimental nuclear physics, high energy physics, nuclear medicine, nuclear tomography, environmental studies and many other fields of use. It is utilized in calorimeters and detectors and also employed as a phosphor in fluoroscopy equipments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-17-5
Molecular Formula CsI
Molecular Weight (g/mol) 259.81
MDL Number MFCD00010962
InChI Key XQPRBTXUXXVTKB-UHFFFAOYSA-M
Synonym cesium iodide, caesium iodide, cesium monoiodide, dicesium diiodide, tricesium triiodide, cesiumiodide, cesium iodide cs2i2, cesium iodide cs3i3, unii-u1p3gvc56l
PubChem CID 24601
IUPAC Name cesium;iodide
SMILES [I-].[Cs+]

Specifications

Boiling Point 1280°C
Melting Point 626°C
Color White
Physical Form Crystalline
Quantity 25 g
Merck Index 14,2014
Solubility Information Soluble in water. Soluble in water and ethanol. Slightly soluble in methanol. Insoluble in acetone.
Formula Weight 259.81
Percent Purity 99.999%
Odor Odorless
Refractive Index 1.7876
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Cesium iodide
Show More Show Less
Hazard Category H315-H317-H319-H335-H361
Hazard Statement GHS H Statement
H302-H315-H319-H317-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 232-145-2
RTECSNumber FLO350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.