Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium nitrate, 99.999% (metals basis)

Catalog No. p-4407371
Click to view available options
Quantity:
25 g
100 g

CAS: 7789-18-6 | CsNO3 | 194.909 g/mol

Cesium nitrate prisms are used in infrared spectroscopy, x-ray phosphors and scintillation counters. It is a precursor material for the synthesis of cesium nitratocuprate and cesium salts. Cesium nitrate doped on carbon-based ruthenium catalyst plays an important role in synthesis of ammonia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-18-6
Molecular Formula CsNO3
Molecular Weight (g/mol) 194.909
MDL Number MFCD00010963
InChI Key NLSCHDZTHVNDCP-UHFFFAOYSA-N
Synonym cesium nitrate, caesium nitrate, nitric acid, cesium salt, cesium i nitrate 1:1, caesium 1+ nitronate, acmc-1bhhg, caesium 1+ ion nitronate, ksc492a7j
PubChem CID 62674
IUPAC Name cesium;nitrate
SMILES [N+](=O)([O-])[O-].[Cs+]

Specifications

Boiling Point (decomposition)
Melting Point 414°C
Color White
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99.999% (Metal basis)
Solubility Information Soluble in water and acetone. Slightly soluble in ethanol.
Formula Weight 194.91
Odor Odorless
Refractive Index 1.55
Sensitivity Hygroscopic
Appearance White crystalline powder
Density 3.68
Chemical Name or Material Cesium nitrate
Show More Show Less
Hazard Category H271-H302-H361f
Precautionary Statement P201-P202-P220-P221-P264-P270-P272-P280-P301+P312-P308+P313-P330-P370+P378q-P370+P380+P375-P372-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: III
EINECSNumber 232-146-8
RTECSNumber FL0700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.