Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium orthovanadate, 99.9% (metals basis)

Catalog No. p-7050652
Click to view available options
Quantity:
10 g
50 g

CAS: 34283-69-7

Cesium orthovanadate, is widely used in chemical research, and also as an intermediate in Pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34283-69-7
MDL Number MFCD00210609
Synonym cesium orthovanadate, tricaesium 1+ vanadate 3-

Specifications

Physical Form Powder
Quantity 10 g
UN Number UN3288
Solubility Information It is soluble in water.
Formula Weight 513.65
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Cesium orthovanadate
Hazard Category H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 251-916-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.