Learn More
Cesium sulfate, Puratronic™, 99.997% (metals basis)
Description
Cesium sulfate is used to prepare dense aqueous solutions for use in isopycnic centrifugation. It is also used for crystal growing purposes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 1,010°C |
Color | White |
Physical Form | Powder |
Quantity | 10 g |
Linear Formula | Cs2SO4 |
Merck Index | 14,2016 |
Solubility Information | Soluble in water (1820g/L). |
Formula Weight | 361.87 |
Percent Purity | 99.997% |
Grade | Puratronic™ |
Show More |
Safety and Handling
Hazard Category | H302-H361f-H373 |
Precautionary Statement | P201-P202-P260-P264b-P270-P281-P301+P312-P308+P313-P330-P501c |
EINECSNumber | 233-662-6 |
RTECSNumber | FL0800000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.