Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CESK1 Antibody (OTI2A5), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272413AF594
Description
CESK1 Monoclonal specifically detects CESK1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
CESK1 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
CCT8L2 | |
Full length recombinant protein of human CESK1 (NP_055221) produced in E.coli. | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI2A5 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
CESK1MGC118839, chaperonin containing TCP1, subunit 8 (theta)-like 2, MGC118840, putative T-complex protein 1 subunit theta-like 2, T-complex protein 1 | |
Mouse | |
0.1 mL | |
150160 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction