Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CESK1 Monoclonal Antibody (1G8)
GREENER_CHOICE

Catalog No. PIMA523095
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA523095 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA523095 Supplier Invitrogen™ Supplier No. MA523095
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MDSTVPSALE LPQRLALNPR ESPRSPEEEE PHLLSSLAAV QTLASVIRPC YGPHGRQKFL VTMKGETVCT GCATAILRAL ELEHPAAWLL REAGQTQAEN SGDGTAFVVL LTEALLEQAE QLLKAGLPRP QLREAYATAT AEVLATLPSL AIQSLGPLED PSWALHSVMN THTLSPMDHL TKLVAHACWA IKELDGSFKP ERVGVCALPG GTLEDSCLLP GLAISGKLCG QMATVLSGAR VALFACPFGP AHPNAPATAR LSSPADLAQF SKGSDQLLEK QVGQLAAAGI NVAVVLGEVD EETLTLADKY GIVVIQARSW MEIIYLSEVL DTPLLPRLLP PQRPGKCQRV YRQELGDGLA VVFEWECTGT PALTVVLRGA TTQGLRSAEQ AVYHGIDAYF QLCQDPRLIP GAGATEMALA KMLSDKGSRL EGPSGPAFLA FAWALKYLPK TLAENAGLAV SDVMAEMSGV HQGGNLLMGV GTEGIINVAQ EGVWDTLIVK AQGFRAVAEV VLQLVTVDEI VVAKKSPTHQ EIWNPDSKKT KKHPPPVETK KILGLNN.

Possible molecular chaperone; assists the folding of proteins upon ATP hydrolysis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CESK1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1G8
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene CCT8L2
Gene Accession No. Q96SF2
Gene Alias CCT8L2; CESK1; chaperonin containing TCP1 subunit 8 like 2; chaperonin containing TCP1, subunit 8 (theta)-like 2; putative T-complex protein 1 subunit theta-like 2; T-complex protein 1 subunit theta-like 2
Gene Symbols CCT8L2
Host Species Mouse
Immunogen CCT8L2 (AAH33797, 1 a.a. ~ 557 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150160
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.