Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CESK1 Monoclonal Antibody (OTI1C1)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526103
Description
CESK1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Possible molecular chaperone; assists the folding of proteins upon ATP hydrolysis.
Specifications
CESK1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96SF2 | |
CCT8L2 | |
Full length human recombinant protein of CCT8L2 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
OTI1C1 | |
Unconjugated | |
CCT8L2 | |
CCT8L2; CESK1; chaperonin containing TCP1 subunit 8 like 2; chaperonin containing TCP1, subunit 8 (theta)-like 2; putative T-complex protein 1 subunit theta-like 2; T-complex protein 1 subunit theta-like 2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
150160 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction