Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CESK1 Monoclonal Antibody (OTI4B10)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526109
Description
CESK1 Monoclonal Antibody for Western Blot, IHC (P), Flow
Possible molecular chaperone; assists the folding of proteins upon ATP hydrolysis.
Specifications
| CESK1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96SF2 | |
| CCT8L2 | |
| Full length human recombinant protein of CCT8L2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI4B10 | |
| Unconjugated | |
| CCT8L2 | |
| CCT8L2; CESK1; chaperonin containing TCP1 subunit 8 like 2; chaperonin containing TCP1, subunit 8 (theta)-like 2; putative T-complex protein 1 subunit theta-like 2; T-complex protein 1 subunit theta-like 2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 150160 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction