Learn More
CETN3 Mouse anti-Human, Clone: OTI4G5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF805822

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene contains four EF-hand calcium binding domains, and is a member of the centrin protein family. Centrins are evolutionarily conserved proteins similar to the CDC31 protein of S. cerevisiae. Yeast CDC31 is located at the centrosome of interphase and mitotic cells, where it plays a fundamental role in centrosome duplication and separation. Multiple forms of the proteins similar to the yeast centrin have been identified in human and other mammalian cells, some of which have been shown to be associated with centrosome fractions. This protein appears to be one of the most abundant centrins associated with centrosome, which suggests a similar function to its yeast counterpart.Specifications
CETN3 | |
Monoclonal | |
Unconjugated | |
CETN3 | |
CDC31, CEN3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
1070 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI4G5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O15182 | |
CETN3 | |
Full length human recombit protein of human CETN3 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.