Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CETP Antibody (2F1), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP315191
This item is not returnable.
View return policy
Description
CETP Monoclonal antibody specifically detects CETP in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
CETP | |
Monoclonal | |
Unconjugated | |
PBS (pH 7.4), 150mM NaCl, and 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry | |
2F1 | |
ELISA, Immunohistochemistry 1:50-1:200 | |
cholesteryl ester transfer protein, cholesteryl ester transfer protein, plasma, HDLCQ10, Lipid transfer protein I | |
A synthesized peptide derived from human CETP. UniProt ID (P11597). | |
100 μg | |
Lipid and Metabolism | |
1071 | |
Store at -20 to -80°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction