Learn More
Invitrogen™ CETP Polyclonal Antibody

Description
PA1-050 detects cholesteryl ester transfer protein (CETP) from human tissues. PA1-050 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~70 kDa band from human liver extracts representing CETP. The PA1-050 immunogen is a synthetic peptide corresponding to residues C D(149) S G R V R T D A P D(159) of human CETP protein.This peptide (Cat. # PEP-209) is available for use in neutralization and control experiments.
Specifications
Specifications
| Antigen | CETP |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1 mg/mL BSA and 0.05% sodium azide |
| Gene | CETP |
| Gene Accession No. | P11597 |
| Gene Alias | BPI fold containing family F; BPIFF; CETP; Cholesteryl ester transfer protein; cholesteryl ester transfer protein plasma; cholesteryl ester transfer protein, plasma; HDLCQ10; Lipid transfer protein I |
| Gene Symbols | CETP |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.