Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CF106 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556950
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556950 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556950 Supplier Invitrogen™ Supplier No. PA556950
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DVTIGEGESI PPDTQFVKTW RIQNSGAEAW PPGVCLKYVG GDQFGHVNMV MVRSLEPQEI ADVSVQMCSP SR Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

Negative regulator of innate antiviral response. Blocks IRF3-dependent cytokine production such as IFNA, IFNB and TNF (PubMed:29802199). Interacts with IRF3 and inhibits IRF3 recruitment to type I IFN promoter sequences while also reducing nuclear levels of the coactivators EP300 and CREBBP (PubMed:29802199). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CF106
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C6orf106
Gene Accession No. Q9H6K1
Gene Alias AU020189; C030010A15; C6orf106; C80239; chromosome 6 open reading frame 106; D17Wsu92e; dJ391O22.4; DNA segment, Chr 17, Wayne State University 92, expressed; FP852; Ilrun; Inflammation and lipid regulator with UBA-like and NBR1-like domains protein; Protein ILRUN; uncharacterized protein C6orf106; uncharacterized protein C6orf106 homolog
Gene Symbols C6orf106
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CF106. Recombinant protein control fragment (Product #RP-104251).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64771
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.