Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CFAP126 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561120
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561120 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561120 Supplier Invitrogen™ Supplier No. PA561120
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NGLCPEILGK PHDPDSQKKL RKKSITKTVQ QARSPTIIPS SPAANLNSPD ELQSSHPSAG HTPGPQRPAK S Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 72%.

CFAP126 acts as a regulator of cilium basal body docking and positioning in mono- and multiciliated cells. Regulates basal body docking and cilia formation in multiciliated lung cells. Regulates kinocilium positioning and stereocilia bundle morphogenesis in the inner ear.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CFAP126
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CFAP126
Gene Accession No. Q5VTH2
Gene Alias 1700009P17Rik; AV282292; C13H1orf192; C1orf192; C38H1orf192; C3H1orf192; C4H1orf192; C5H1orf192; CFAP126; cilia and flagella associated protein 126; cilia- and flagella-associated protein 126; Flattop; Fltp; hypothetical protein LOC510399; protein Flattop; RGD1562658; UPF0740 protein C1orf192; UPF0740 protein C1orf192 homolog
Gene Symbols CFAP126
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CFAP126. Recombinant protein control fragment (Product #RP-94537).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 257177
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.