Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CFAP299 Antibody (OTI1A6), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272466AF647
Description
CFAP299 Monoclonal specifically detects CFAP299 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| CFAP299 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CFAP299 | |
| Full length recombinant protein of human C4orf22 (NP_689983) produced in HEK293T cells. | |
| 0.1 mL | |
| 255119.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI1A6 | |
| Western Blot, Immunohistochemistry | |
| chromosome 4 open reading frame 22, hypothetical protein LOC255119, MGC35043 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction