Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CFAP299 Antibody (OTI1A6), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272466FR
Description
CFAP299 Monoclonal specifically detects CFAP299 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CFAP299 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
CFAP299 | |
Full length recombinant protein of human C4orf22 (NP_689983) produced in HEK293T cells. | |
0.1 mL | |
255119.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI1A6 | |
Western Blot, Immunohistochemistry | |
chromosome 4 open reading frame 22, hypothetical protein LOC255119, MGC35043 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction