Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CFAP61 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552649
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552649 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552649 Supplier Invitrogen™ Supplier No. PA552649
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HMKFNNLTLI STHGLPGKKL LDTEQRKFLA SDHCFNDKDY ALMSLCSWVN VVVGRMTGID RAAKHVVLST DEIVPYDHLI LCTGQQYQVP CPTEADISQH LTNREVPNSS QRRYTGKVPC NHFTLNEEED CFKALIWIRN NSITTE Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 17%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CFAP61
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CFAP61
Gene Accession No. Q8NHU2
Gene Alias 4930529M08Rik; BB178539; BC024760; C20orf26; CaM-IP3; CFAP61; cilia and flagella associated protein 61; cilia- and flagella-associated protein 61; dJ1002M8.3; dJ1178H5.4; RGD1311519
Gene Symbols CFAP61
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CFAP61. Recombinant protein control fragment (Product #RP-90022).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26074
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.