Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CFAP99 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145010
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145010 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145010 Supplier Invitrogen™ Supplier No. PA5145010
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLQFPPRIRK TPKLTFYRPD NILVKLNTTA ILREGALYQR QVEQELQRVD KLVDGAGDFS EFFEWQKKMQ Antigen sequence identity: Mouse 77%; Rat 77%.

CFAP99 (Cilia and Flagella Associated Protein 99) is a protein that is primarily expressed in the cilia and flagella of cells. It is involved in the regulation of ciliary and flagellar motility, and mutations in the CFAP99 gene have been linked to primary ciliary dyskinesia, a genetic disorder that affects the function of cilia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CFAP99
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CFAP99
Gene Accession No. D6REC4
Gene Alias Cilia- and flagella-associated protein 99
Gene Symbols CFAP99
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 272-341. Recombinant protein control fragment (Product #RP-109843).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 402160
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.