Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CFC1B/CFC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575660
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Cryptic is a 224 amino acid member of the epidermal growth factor-Cripto-1/ FRL1/cryptic family that consists of extracellular and cell-associated proteins which function as co-receptors for the transforming growth factor b subfamily of proteins. Cryptic is involved in the appropriate establishment of the left-right axis of the organism, and it may play a role in the development of mesoderm and/or neural patterning during gastrulation. Loss-of-function mutations in CFC1, the gene encoding for Cryptic, are associated with human left-right laterality defects. Defects in CFC1 also cause visceral heterotaxy (HTX2), an autosomal disease characterized by a variable group of congenital anomalies that include complex cardiac malformations such as conotruncal heart malformations that may lead to cardiac outflow tract defects, pulmonary atresia, double-outlet right ventricle, truncus arteriosus communis and aortic arch anomalies.
Specifications
CFC1B/CFC1 | |
Polyclonal | |
Unconjugated | |
CFC1B | |
AV265756; b2b970Clo; CFC1; CFC1B; cripto, FRL-1, cryptic family 1; cripto, FRL-1, cryptic family 1B; cryptic; Cryptic family protein 1; cryptic family protein 1B; Cryptic protein; DTGA2; heterotaxy 2 (autosomal dominant); HTX2; RGD1562188 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12627, 501121, 55997, 653275 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P0CG36, P0CG37, P97766 | |
CFC1, CFC1B | |
Synthetic peptide corresponding to residues in Human CFC1B/CFC1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction