Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CFDP1 Mouse anti-Human, Unconjugated, Clone: 15E5.1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABN39625UL
Description
Anti-CFDP1, clone 15E5.1, Cat. No. MABN396, is a highly specific mouse monoclonal antibody that targets CFDP1 and has been tested for use in Western Blotting.
Specifications
CFDP1 | |
Monoclonal | |
Please refer to lot specific datasheet. | |
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
CFDP1;BCNT;CENP-29 | |
GST-taged recombinant fragment corresponding to the first 111 amino acids from the N-terminal region. | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Antibodies | |
IgG1 κ |
Western Blot | |
15E5.1 | |
Unconjugated | |
Bucentaur;CFDP1 | |
Mouse | |
∽45 kDa observed; 33.59 kDa calculated. Uncharacterized bands may be observed in some lysate(s). | |
25 μL | |
Neuroscience | |
Clone 15E5.1 detects Craniofacial development protein 1 in human. It targets an epitope within the first 111 amino acids from the N-terminal half. | |
Stable for 1 year at 2-8°C from date of receipt. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction