Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CFDP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534953
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534953 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534953 Supplier Invitrogen™ Supplier No. PA534953
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, HeLa, HepG2, A375. Predicted reactivity: Dog (91%), Rhesus Monkey (98%), Bovine (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

May play a role during embryogenesis. Its mouse homolog is involved in embryogenesis and craniofacial development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CFDP1
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene CFDP1
Gene Accession No. O88271, Q9UEE9
Gene Alias 27 kDa craniofacial protein; AA408409; Bcnt; Bucentaur; CENP-29; centromere protein 29; Cfdp; CFDP1; CFDP1 protein; Cp27; Craniofacial development protein 1; fi15f05; h-type BCNT protein; human-type Bcnt; p97; p97 homologous protein; phosphoprotein (Bucentaur); Protein Cp27; si:dkey-6e12.6; SWC5; Unknown (protein for MGC:136405); Unknown (protein for MGC:137094); wu:fi15f05; Yeti; zgc:136405
Gene Symbols CFDP1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human CFDP1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10428, 23837
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.