Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CFDP1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982171
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2171 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2171 Supplier Bioss Supplier No. BS12357R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May play a role during embryogenesis. Its mouse homolog is involved in embryogenesis and craniofacial development.

Specifications

Antigen CFDP1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CFDP1
Gene Accession No. Q9UEE9
Gene Alias 27 kDa craniofacial protein; AA408409; Bcnt; BOS_16922; Bucentaur; CENP-29; centromere protein 29; Cfdp; Cfdp1; CFDP1 protein; Cp27; Craniofacial development protein 1; fi15f05; h-type BCNT protein; human-type Bcnt; LOC100350560; P97; p97 homologous protein; phosphoprotein (Bucentaur); Protein Cp27; RGD motif, leucine rich repeats, tropomodulin domain and proline-rich containing; rltpr; si:dkey-6e12.6; SWC5; Unknown (protein for MGC:136405); wu:fi15f05; Yeti; zgc:136405
Gene Symbols CFDP1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CFDP1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10428
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.