Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CFHR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102655
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102655 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102655 Supplier Invitrogen™ Supplier No. PA5102655
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CFHR1.

This gene encodes a secreted protein belonging to the complement factor H protein family. It binds to Pseudomonas aeruginosa elongation factor Tuf together with plasminogen, which is proteolytically activated. It is proposed that Tuf acts as a virulence factor by acquiring host proteins to the pathogen surface, controlling complement, and facilitating tissue invasion. Mutations in this gene are associated with an increased risk of atypical hemolytic-uremic syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CFHR1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CFHR1
Gene Accession No. Q03591
Gene Alias AI194696; CFHL; Cfhl1; CFHL1P; CFHR1; CFHR1P; CFHRB; complement component factor h-like 1; complement factor H related 1; complement factor H-related 1; complement factor H-related 1 pseudogene; complement factor H-related protein; complement factor H-related protein 1; FHR1; FHR-1; H factor (complement)-like 1; H factor (complement)-like 2; h factor-like protein 1; H36; H36-1; H36-2; H-factor-like 1; HFL1; HFL2
Gene Symbols CFHR1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CFHR1(Accession Q03591), corresponding to amino acid residues E274-Y324.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 289057, 3078, 50702
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.