Learn More
Invitrogen™ CFHR1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120829
Description
Positive test controls include: LO2, HeLa, NCI-H460, Mouse liver, Mouse lung, Mouse heart, Rat liver. The target is usually found in the following locations: Secreted. Immunogen sequence: PTVQNAHILS RQMSKYPSGE RVRYECRSPY EMFGDEEVMC LNGNWTEPPQ CKDSTGKCGP PPPIDNGDIT SFPLSVYAPA SSVEYQCQNL YQLEGNKRIT CRNGQWSEPP KCLHPCVISR EIMENYNIAL RWTAKQKLYL RTGESAEFVC KRGYRLSSRS HTLRTTCWDG KLEYPTCAKR.
This gene encodes a secreted protein belonging to the complement factor H protein family. It binds to Pseudomonas aeruginosa elongation factor Tuf together with plasminogen, which is proteolytically activated. It is proposed that Tuf acts as a virulence factor by acquiring host proteins to the pathogen surface, controlling complement, and facilitating tissue invasion. Mutations in this gene are associated with an increased risk of atypical hemolytic-uremic syndrome.
Specifications
CFHR1 | |
Polyclonal | |
Unconjugated | |
CFHR1 | |
AI194696; CFHL; Cfhl1; CFHL1P; CFHR1; CFHR1P; CFHRB; complement component factor h-like 1; complement factor H related 1; complement factor H-related 1; complement factor H-related 1 pseudogene; complement factor H-related protein; complement factor H-related protein 1; FHR1; FHR-1; H factor (complement)-like 1; H factor (complement)-like 2; h factor-like protein 1; H36; H36-1; H36-2; H-factor-like 1; HFL1; HFL2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
289057, 3078, 50702 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
3.08 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q03591 | |
CFHR1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 151-330 of human CFHR1 (NP_002104.2). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.