Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CFHR2 Monoclonal Antibody (01)
GREENER_CHOICE

Catalog No. PIMA530230
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530230 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530230 Supplier Invitrogen™ Supplier No. MA530230
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human CFHR2.

Might be involved in complement regulation. Can associate with lipoproteins and may play a role in lipid metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CFHR2
Applications ELISA
Classification Monoclonal
Clone 01
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CFHR2
Gene Accession No. P36980
Gene Alias CFHL2; CFHR2; complement factor H related 2; complement factor H-related 2; complement factor H-related protein 2; DDESK59; factor H-related gene 2; FHR2; FHR-2; H factor (complement)-like 3; h factor-like 3; H factor-like protein 2; HFL3
Gene Symbols CFHR2
Host Species Mouse
Immunogen Recombinant Human CFHR2 protein (Met1-Lys270).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3080
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.