Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CFHR3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596801
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596801 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596801 Supplier Invitrogen™ Supplier No. PA596801
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AVPCLRKCYF PYLENGYNQN YGRKFVQGNS TEVACHPGYG LPKAQTTVTC TEKGWSPTPR CIRVRTCSKS DIEIENGFIS ESSSIYILNK EIQYKCKPGY ATADGNSSGS ITCLQNGWSA Q; Positive Samples: A-549, HepG2, NIH/3T3, 293T, Mouse liver, Rat liver.

The protein encoded by this gene is a secreted protein, which belongs to the complement factor H-related protein family. It binds to heparin, and may be involved in complement regulation. Mutations in this gene are associated with decreased risk of age-related macular degeneration, and with an increased risk of atypical hemolytic-uremic syndrome. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CFHR3
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.79 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CFHR3
Gene Accession No. Q02985
Gene Alias CFHL3; CFHR3; complement factor H related 3; complement factor H-related 3; complement factor H-related protein 3; DOWN16; EG624286; FHR3; FHR-3; H factor-like 4; H factor-like protein 3; HLF4
Gene Symbols CFHR3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 80-200 of human CFHR3 (NP_0663032).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10878, 624286
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.