Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CFHR3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112384
Description
CFHR3 Polyclonal Antibody for Western Blot, ELISA
The protein encoded by this gene is a secreted protein, which belongs to the complement factor H-related protein family. It binds to heparin, and may be involved in complement regulation. Mutations in this gene are associated with decreased risk of age-related macular degeneration, and with an increased risk of atypical hemolytic-uremic syndrome. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| CFHR3 | |
| Polyclonal | |
| Unconjugated | |
| CFHR3 | |
| CFHL3; CFHR3; complement factor H related 3; complement factor H-related 3; complement factor H-related protein 3; DOWN16; EG624286; FHR3; FHR-3; H factor-like 4; H factor-like protein 3; HLF4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10878 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 0.57 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q02985 | |
| CFHR3 | |
| Recombinant Human Complement factor H-related protein 3 protein (80-200AA). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction