Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CFHR4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114239
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114239 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114239 Supplier Invitrogen™ Supplier No. PA5114239
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SNGEWSEPPR CIHPCIITEE NMNKNNIQLK GKSDIKYYAK TGDTIEFMCK For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%.

This gene is a member of the complement factor H (CFH) gene family, and encodes one of the 5 CFH-related (CFHR) proteins. These 5 genes are closely linked to the CFH gene on chromosome 1q31-q32. The CFHRs are secreted plasma proteins synthesized primarily by the hepatocytes, and composed of highly-related short consensus repeats (SCRs). This protein enhances the cofactor activity of CFH, and is involved in complement regulation. It can associate with lipoproteins and may play a role in lipid metabolism. Alternatively spliced transcript variants encoding different isoforms (varying in the number of SCRs) have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CFHR4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CFHR4
Gene Accession No. Q92496
Gene Alias CFHL4; CFHR4; complement factor H; complement factor H related 4; complement factor H-related 4; complement factor H-related protein 4; FHR4; FHR-4
Gene Symbols CFHR4
Host Species Rabbit
Immunogen Immunogen sequence: SNGEWSEPPR CIHPCIITEE NMNKNNIQLK GKSDIKYYAK TGDTIEFMCK.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10877
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.