Learn More
Invitrogen™ CFHR4 Polyclonal Antibody

Description
Peptide sequence: LWVSCANGQE VKPCDFPEIQ HGGLYYKSLR RLYFPAAAGQ SYSYYCDQNF Sequence homology: Human: 100%; Rat: 79%.
Specifications
Specifications
| Antigen | CFHR4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | CFHR4 |
| Gene Accession No. | Q92496 |
| Gene Alias | CFHL4; CFHR4; complement factor H; complement factor H related 4; complement factor H-related 4; complement factor H-related protein 4; FHR4; FHR-4 |
| Gene Symbols | CFHR4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.