Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CFHR5 Mouse anti-Human, Mouse, Clone:3A10A5, Abnova™
Mouse monoclonal antibody raised against recombinant human CFHR5.
Supplier: Abnova Corporation MAB17205
Specifications
CFHR5 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Immunocytochemistry (1:200-1:1000) Flow Cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. |
|
CFHR5 | |
Mouse | |
100 μg | |
Human, Mouse | |
Antibody | |
IgG2a |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
3A10A5 | |
complement factor H-related 5 | |
In PBS (0.05% sodium azide) | |
CFHL5, FHR-5, FHR5, FLJ10549, MGC133240 | |
Recombinant protein corresponding to amino acid 344-569 of human CFHR5 from E. coli. | |
81494 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction