Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CFI Monoclonal Antibody (OTI4A8), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA806031

Description
Factor I, a serine protease, is a regulatory component of both the alternative and the classical complement pathway. Factor I is synthesized as a single chain precursor of 88 kDa, processed to yield the acitve form, composed of two disulfide-linked chains, the a-chain (50 kDa) and the beta-chain (38 kDa). Plasma concentration is 35 ug/ml.Specifications
CFI | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P05156 | |
CFI | |
Human recombit protein fragment corresponding to amino acids 340-583 of human CFI produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI4A8 | |
Unconjugated | |
CFI | |
AHUS3, ARMD13, C3BINA, C3b-INA, FI, IF, KAF | |
Mouse | |
Affinity Chromatography | |
RUO | |
3426 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction