Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CFI Mouse anti-Human, Clone: OTI2B5, lyophilized, TrueMAB™
SDP

Catalog No. 501667701
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-7701 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-7701 Supplier OriGene Supplier No. CF806115
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

Factor I, a serine protease, is a regulatory component of both the alternative and the classical complement pathway. Factor I is synthesized as a single chain precursor of 88 kDa, processed to yield the acitve form, composed of two disulfide-linked chains, the a-chain (50 kDa) and the beta-chain (38 kDa). Plasma concentration is 35 ug/ml.

Specifications

Antigen CFI
Applications Western Blot
Classification Monoclonal
Clone OTI2B5
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene CFI
Gene Accession No. P05156
Gene Alias AHUS3, ARMD13, C3BINA, C3b-INA, FI, IF, KAF
Gene Symbols CFI
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 340-583 of human CFI produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3426
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.