Learn More
CFI Mouse anti-Human, Clone: OTI7D6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF805922

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Factor I, a serine protease, is a regulatory component of both the alternative and the classical complement pathway. Factor I is synthesized as a single chain precursor of 88 kDa, processed to yield the acitve form, composed of two disulfide-linked chains, the a-chain (50 kDa) and the beta-chain (38 kDa). Plasma concentration is 35 ug/ml.Specifications
CFI | |
Monoclonal | |
Unconjugated | |
CFI | |
AHUS3, ARMD13, C3BINA, C3b-INA, FI, IF, KAF | |
Mouse | |
Affinity Chromatography | |
RUO | |
3426 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI7D6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P05156 | |
CFI | |
Human recombit protein fragment corresponding to amino acids 340-583 of human CFI produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.