Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD CFSE

Catalog No. bdb565082 Shop All BD Biosciences Products
Click to view available options
Quantity:
1 mg

Catalog No. BDB565082

Supplier: BD 565082

Only null left
Add to Cart
Add to Cart

Used for flow cytometric monitoring of cell divisions. BD Horizon™ CFSE is non-fluorescent dye passively diffuses across cell membranes and is cleaved by intracellular esterases within viable cells.

  • Blue laser excitable dye
  • Used for flow cytometric monitoring of cell divisions
  • The non-fluorescent dye passively diffuses across cell membranes and is cleaved by intracellular esterases within viable cells
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
For Use With (Application) Intracellular staining (flow cytometry)
Color Blue
Storage Requirements ≤ -20°C
Stain Type Laser excitable dye
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.