Learn More
Invitrogen™ eBioscience™ CFSE
Description
- Molecular weight of 557.47g/mol
- After the acetate groups are cleaved, it has a peak excitation of 494nm and peak emission of 521nm
- Each vial may be reconstituted to stock concentration of 10 mM with 90μL of anhydrous DMSO
- Once reconstituted it should be used within 6 months and protected from light and stored at −20°C with dessicant; avoid freeze-thawing
- Unit Size: 5 x 500μg
Specifications
Specifications
| Detection Method | Fluorescence |
| For Use With (Equipment) | Fluorescence Microscope, Flow Cytometer |
| Product Type | CFSE |
| Format | 5 x 500 μg |
| Product Line | eBioscience |
| Quantity | 5 x 500 μg |
| Emission Wavelength | 494/521 |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.