Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CFTR, anti-Human, Clone: CFTR/1775R, Abnova™

Catalog No. 89035196 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-196 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-196 Supplier Abnova Corporation Supplier No. RAB00317
Add to Cart
Add to Cart

Monoclonal Antibody

Specifications

Antigen CFTR
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CFTR/1775R
Conjugate Unconjugated
Dilution Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Gene cystic fibrosis transmembrane conductance regulator (ATP-binding cassette sub-family C, member 7)
Gene Alias ABC35/ABCC7/CF/CFTR/MRP/MRP7/TNR-CFTR/dJ760C5.1
Gene Symbols CFTR
Immunogen Original antibody is raised against recombinant protein corresponding to amino acids 258-385 of human CFTR.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1080
Target Species Human
Content And Storage Store at 4°C.
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.