Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CFTR Antibody (CF3), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$665.50

Specifications

Antigen CFTR
Clone CF3
Dilution Western Blot 1:500, Flow Cytometry 1:100, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:200, Blocking/Neutralizing 50 - 200 molar excess
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB300511
SDP
View Documents
Novus Biologicals
NB300511
100 μL
Each of 1 for $665.50
Only null left
Add to Cart
 
Description

Description

CFTR Monoclonal antibody specifically detects CFTR in Human, Mouse, Rat, Chicken, Golden Syrian Hamster samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Block/Neutralize.
Specifications

Specifications

CFTR
Western Blot 1:500, Flow Cytometry 1:100, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:200, Blocking/Neutralizing 50 - 200 molar excess
Monoclonal
Ascites
RUO
P13569
1080
Synthetic Peptide: G(103) R I I A S Y D P D N K E E R(117)
Primary
Store at -20C. Avoid freeze-thaw cycles.
168 kDa
CF3
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Unconjugated
Mouse
ABC Transporters, Cancer, Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones
ABC35, ABCC7CF, ATP-binding cassette sub-family C member 7, ATP-binding cassette transporter sub-family C member 7, cAMP-dependent chloride channel, CFTR/MRP, Channel conductance-controlling ATPase, cystic fibrosis transmembrane conductance regulator, cystic fibrosis transmembrane conductance regulator (ATP-binding cassettesub-family C, member 7), cystic fibrosis transmembrane conductance regulator, ATP-binding cassette(sub-family C, member 7), dJ760C5.1, EC 3.6.3, MRP7EC 3.6.3.49, TNR-CFTR
CFTR
IgM
Unpurified
CFTR (CF3)
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.