Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CFTR Monoclonal Antibody (L12B4), Invitrogen™

Catalog No. PIMA511763
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA511763 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA511763 Supplier Thermo Scientific Supplier No. MA511763

Mouse Monoclonal Antibody

MA5-11763 targets Cystic Fibrosis Transmembrane Regulator in IF, IP, and WB applications and shows reactivity with Human, mouse, and Rat samples. The MA5-11763 immunogen is recombinant protein encoding NBF1 domain of human CFTR.

CFTR Monoclonal antibody specifically detects CFTR in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CFTR
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone L12B4
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene CFTR
Gene Accession No. P13569, P26361, P34158
Gene Alias ABC35; Abcc7; ATP Binding Cassette Superfamily C Member 7 (ABCC7); ATP-binding cassette sub-family C member 7; ATP-binding cassette transporter sub-family C member 7; ATP-binding cassette, subfamily c, member 7; AW495489; cAMP-dependent chloride channel; CF; CFTR; CFTR/MRP; Channel conductance-controlling ATPase; cystic fibrosis transmembrane conductance regulator; cystic fibrosis transmembrane conductance regulator (ATP-binding cassette sub-family C, member 7); cystic fibrosis transmembrane conductance regulator homolog; cystic fibrosis transmembrane conductance regulator homolog; ATP-binding cassette, subfamily c, member 7; dJ760C5.1; MRP7; RGD1561193; tcag7.78; TNR CFTR; TNR-CFTR
Gene Symbols CFTR
Host Species Mouse
Immunogen Recombinant protein encoding NBF1 domain of human CFTR
Purification Method Protein G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1080, 12638, 24255
Target Species Human, Mouse, Rat
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.