Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CFTR Mouse anti-Human, Mouse, Clone-CFTR/1342, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254505200UG
Description
Human chromosome location: 7q31.2 IHC-P:Staining of formalin-fixed tissues is enhanced by boiling tissue sections in 10mM Tris with 1mM EDTA pH 9.0 for 10-20 min followed by cooling at RT for 20 minuteSpecifications
CFTR | |
Monoclonal | |
1 mg/mL | |
Western Blot 0.5 - 1 ug/ml, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml | |
ABC35, ABCC7CF, ATP-binding cassette sub-family C member 7, ATP-binding cassette transporter sub-family C member 7, cAMP-dependent chloride channel, CFTR/MRP, Channel conductance-controlling ATPase, cystic fibrosis transmembrane conductance regulator, cystic fibrosis transmembrane conductance regulator (ATP-binding cassettesub-family C, member 7), cystic fibrosis transmembrane conductance regulator, ATP-binding cassette(sub-family C, member 7), dJ760C5.1, EC 3.6.3, MRP7EC 3.6.3.49, TNR-CFTR | |
Mouse | |
167 kDa | |
200 μg | |
ABC Transporters, Cancer, Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones | |
1080 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
CFTR/1342 | |
Unconjugated | |
PBS with No Preservative | |
CFTR | |
Recombinant human full length CFTR | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction