Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CFTR Mouse anti-Human, Unconjugated, Clone: rCFTR/1342, Abnova™

Catalog No. 89337892 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-892 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-892 Supplier Abnova Corporation Supplier No. RAB00467
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

CFTR Monoclonal antibody specifically detects CFTR in Human samples. It is validated for WB, IHC-P, IF.

Specifications

Antigen CFTR
Applications Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone rCFTR/1342
Conjugate Unconjugated
Description cystic fibrosis transmembrane conductance regulator (ATP-binding cassette sub-family C, member 7)
Gene CFTR
Gene Alias ABC35, ABCC7, CF, CFTR/MRP, MRP7, TNR-CFTR, dJ760C5.1
Host Species Mouse
Immunogen Original antibody is raised against recombinant protein corresponding to full length human CFTR
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 1080
Target Species Human
Content And Storage Store at 4°C
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.