Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CFTRinh 172
GREENER_CHOICESDP

Catalog No. 343010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-301-0 10 mg
34-305-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 34-301-0 Supplier Tocris Bioscience™ Supplier No. 3430/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Voltage-independent, selective CFTR chloride channel blocker

Voltage-independent, selective CFTR chloride channel blocker (Ki = 300 nM) that alters channel gating. Blocks intestinal fluid secretion induced by cholera toxin and Escherichia coli and suppresses cyst growth in animal models of polycystic kidney disease. Orally active. Inhibits mitochondrial respiration and increases reactive oxygen species (ROS) production independently of CFTR in several cell lines.

Chemical Identifiers

CAS 307510-92-5
Molecular Formula C18H10F3NO3S2

Specifications

Quantity 10 mg
Formula Weight 409.4
Purity >98%
Molecular Weight (g/mol) 409.397
Chemical Name or Material CFTRinh 172
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.