Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CG2727 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5144082
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5144082 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5144082 Supplier Invitrogen™ Supplier No. PA5144082
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control - WB: normal drosophila melanogaster head tissues, CG2727 KO / +, normal drosophila melanogaster thorax tissues, CG2727 KO / +.

CG2727 is a gene in fruit flies that encodes a protein involved in the regulation of gene expression. Specifically, it has been shown to play a role in the regulation of transcription, the process by which genetic information stored in DNA is used to produce RNA molecules. This gene has also been linked to the development of the nervous system and the maintenance of cell polarity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CG2727
Applications Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene emp
Gene Accession No. Q92626
Gene Alias Dmel; Emp
Gene Symbols emp
Host Species Rabbit
Immunogen Drosophila CG2727 between 71-289.
Purification Method Antigen affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 37999
Target Species Drosophila
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.