Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cGAS Rabbit anti-Human, Mouse, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABF124MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABF124MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABF124MI Supplier MilliporeSigma Supplier No. ABF124
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-cGAS antibody is validated for use in Western Blotting for the detection of cGAS.
TRUSTED_SUSTAINABILITY

Specifications

Antigen cGAS
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Cyclic GMP-AMP synthase;cGAMP synthase;cGAS;h-cGAS;Mab-21 domain-containing protein 1
Gene Symbols MB21D1;C6orf150
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human cGAS near the C-terminus.
Molecular Weight of Antigen ∽54 kDa observed
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_612450
Target Species Human, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.