Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CGBP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111200
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111200 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111200 Supplier Invitrogen™ Supplier No. PA5111200
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VKVKHVKRRE KKSEKKKEER YKRHRQKQKH KDKWKHPERA DAKDPASLPQ CLGPGCVRPA QPSSKYCSDD CGMKLAANRI YEILPQRIQQ WQQSPCIAEE HGKKLLERIR REQQSARTRL QEMERRFHEL EAIILRAKQQ Highest antigen sequence indentity to the following orthologs: Rat - 99%, Mouse - 99%.

Proteins that contain a CXXC motif within their DNA-binding domain, such as CXXC1, recognize CpG sequences and regulate gene expression (Carlone and Skalnik, 2001).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CGBP
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CXXC1
Gene Accession No. Q9P0U4
Gene Alias 2410002I16Rik; 5830420C16Rik; AI426635; CFP 1; CFP1; CGBP; CpG binding protein; cpG-binding protein; cpG-binding protein; CXXC-type zinc finger protein 1; CXXC 1; CXXC finger 1; CXXC finger 1 (PHD domain); CXXC finger protein 1; Cxxc1; CXXC-type zinc finger protein 1; DNA-binding protein with PHD finger and CXXC domain; hCGBP; HsT2645; PCCX 1; Pccx1; PHD finger and CXXC domain-containing protein 1; PHF 18; PHF18; SPP 1; SPP1; ZCGPC 1; ZCGPC1; zinc finger, CpG binding-type containing 1
Gene Symbols CXXC1
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human CGBP. Recombinant protein control fragment (Product #RP-89370).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30827
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.