Learn More
Invitrogen™ CGBP Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120214
Description
Positive test controls include: HepG2, BT-474, HepG2, Mouse thymus. The target is usually found in the following locations: Nucleus speckle. Immunogen sequence: MEGDGSDPEP PDAGEDSKSE NGENAPIYCI CRKPDINCFM IGCDNCNEWF HGDCIRITEK MAKAIREWYC RECREKDPKL EIRYRHKKSR ERDGNERDSS EPRDEGGGRK RPVPDPDLQR RAGSGTGVGA MLARGSASPH KSSPQPLVAT PSQHHQQQQQ QIKRSARMCG ECEACRRTED CGHCDFCRDM KKFGGPNKIR QKCRLRQCQL.
Proteins that contain a CXXC motif within their DNA-binding domain, such as CXXC1, recognize CpG sequences and regulate gene expression (Carlone and Skalnik, 2001).
Specifications
| CGBP | |
| Polyclonal | |
| Unconjugated | |
| CXXC1 | |
| 2410002I16Rik; 5830420C16Rik; AI426635; CFP 1; CFP1; CGBP; CpG binding protein; cpG-binding protein; cpG-binding protein; CXXC-type zinc finger protein 1; CXXC 1; CXXC finger 1; CXXC finger 1 (PHD domain); CXXC finger protein 1; Cxxc1; CXXC-type zinc finger protein 1; DNA-binding protein with PHD finger and CXXC domain; hCGBP; HsT2645; PCCX 1; Pccx1; PHD finger and CXXC domain-containing protein 1; PHF 18; PHF18; SPP 1; SPP1; ZCGPC 1; ZCGPC1; zinc finger, CpG binding-type containing 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 30827, 74322 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot | |
| 0.72 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9CWW7, Q9P0U4 | |
| CXXC1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human CXXC1 (NP_001095124.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.