Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CGI-09 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | CGI-09 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
CGI-09 Polyclonal specifically detects CGI-09 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
CGI-09 | |
Polyclonal | |
Rabbit | |
Human | |
51605 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LKDKGIKGEEIVQQLIENSTTFRDKTEFAQDKYIKKKKKKYEAIITVVKPSTRILSIMYYAREPGKINHMRYDTLAQMLTLGNIRAGN | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
CGI-09, GCD10, Gcd10p, KIAA1153, MGC5029, TRM6, tRNA (adenine-N(1)-)-methyltransferase non-catalytic subunit TRM6, tRNA methyltransferase 6 homolog (S. cerevisiae), tRNA(m1A58)-methyltransferase subunit TRM6, tRNA(m1A58)MTase subunit TRM6 | |
TRMT6 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title