Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CGI-09 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP150039
Description
CGI-09 Polyclonal specifically detects CGI-09 in Human samples. It is validated for Immunoprecipitation.Specifications
CGI-09 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
TRMT6 | |
The immunogen for this product maps to a region between residue 325 and 375 of human tRNA Methyltransferase 6 using the numbering given in entry NP_057023.2 (GeneID 51605). | |
100 μL | |
Primary | |
Human | |
IgG |
Immunoprecipitation | |
1.0 mg/mL | |
Immunoprecipitation 2-5 mcg/mg | |
CGI-09, GCD10, Gcd10p, KIAA1153, MGC5029, TRM6, tRNA (adenine-N(1)-)-methyltransferase non-catalytic subunit TRM6, tRNA methyltransferase 6 homolog (S. cerevisiae), tRNA(m1A58)-methyltransferase subunit TRM6, tRNA(m1A58)MTase subunit TRM6 | |
Rabbit | |
Affinity Purified | |
RUO | |
51605 | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction