Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CGI 62 Antibody (OTI4C1), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271896MFV450
Description
CGI 62 Monoclonal antibody specifically detects CGI 62 in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
CGI 62 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
51101 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
OTI4C1 | |
Flow Cytometry | |
C8orf70, CGI-62, chromosome 8 open reading frame 70, family with sequence similarity 164, member A, hypothetical protein LOC51101 | |
Full length human recombinant protein of human FAM164A(NP_057094) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction