Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CGI 62 Antibody (OTI4C1), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271896PE
Description
CGI 62 Monoclonal specifically detects CGI 62 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
| CGI 62 | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| ZC2HC1A | |
| Full length recombinant protein of human CGI 62 (NP_057094) produced in E.coli. | |
| 0.1 mL | |
| 51101.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI4C1 | |
| Flow Cytometry | |
| C8orf70, CGI-62, chromosome 8 open reading frame 70, family with sequence similarity 164, member A, hypothetical protein LOC51101 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction